Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0852

Sigma-Aldrich

CC4

≥98% (HPLC)

Synonym(s):

(1R,5S)-1,2,3,4,5,6-Hexahydro-3-[2-[(1R,5S)-1,5,6,8-tetrahydro-8-oxo-1,5-methano-2H-pyrido[1,2-a][1,5]diazocin-3(4H)-yl]ethyl]-1,5-Methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, 1,2-BisN-cytisinylethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30N4O2
CAS Number:
Molecular Weight:
406.52
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C24H30N4O2/c29-23-5-1-3-21-19-9-17(13-27(21)23)11-25(15-19)7-8-26-12-18-10-20(16-26)22-4-2-6-24(30)28(22)14-18/h1-6,17-20H,7-16H2/t17-,18-,19+,20+/m0/s1

InChI key

LEXDAVFCJPDCNA-VNTMZGSJSA-N

Biochem/physiol Actions

CC4 is a a partial agonist of α4β2 and α6β2 nicotinic acetylcholine receptors with low affinity for α3β4 and α7 containing receptors. CC4 reduces nicotine-induced self-administration and conditioned place preference without affecting motor functions.
CC4 is a a partial agonist of α4β2 and α6β2 nicotinic acetylcholine receptors.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sigma-Aldrich offers many products related to nicotinic acetylcholine receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service