Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0840

Sigma-Aldrich

Staphyloccocus UPPS Inhibitor methanesulfonate

≥98% (HPLC)

Synonym(s):

N4,N4′-bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]-[1,1′-Biphenyl]-4,4′-dicarboxamide methanesulfonate salt, NSC 50460

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$371.00

$371.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$371.00

About This Item

Empirical Formula (Hill Notation):
C32H28N6O2 · 2CH3SO3H
CAS Number:
Molecular Weight:
720.82
UNSPSC Code:
51111800
NACRES:
NA.77

$371.00


Check Cart for Availability

Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Staphyloccocus UPPS Inhibitor methanesulfonate is a potent inhibitor of the bacteria enzyme undecaprenyl diphosphate synthase (UPPS).
Staphyloccocus UPPS Inhibitor methanesulfonate is a potent inhibitor of the bacteria enzyme undecaprenyl diphosphate synthase (UPPS). Staphyloccocus UPPS Inhibitor kills both wild type and methicillin-resitant strains of Staphylococcus aureus (MRSA). Staphyloccocus UPPS Inhibitor protects mice from death after infection with MRSA.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service