Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0820

Sigma-Aldrich

HJC0350

≥98% (HPLC)

Synonym(s):

2,4-Dimethyl-1-[(2,4,6-trimethylphenyl)sulfonyl]-1H-pyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19NO2S
CAS Number:
Molecular Weight:
277.38
UNSPSC Code:
41106305
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

storage temp.

room temp

InChI

1S/C15H19NO2S/c1-10-6-12(3)15(13(4)7-10)19(17,18)16-9-11(2)8-14(16)5/h6-9H,1-5H3

InChI key

AFZWZVLPIMHLSE-UHFFFAOYSA-N

Biochem/physiol Actions

HJC0350 is a potent inhibitor of the Exchange Protein directly Activated by cAMP (EPAC) family member EPAC2.
HJC0350 is a potent inhibitor of the Exchange Protein directly Activated by cAMP (EPAC) family member EPAC2. HJC0350 blocks EPAC2 in enzymatic and cell-based assays, with no detectable acitivity against EPAC1, or protein kinase A (PKA).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service