Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0799

Sigma-Aldrich

CYM 50179

≥98% (HPLC)

Synonym(s):

2,6-Dibromo-3-[2-(2,4-dichlorophenoxy)ethoxy]-pyridine, CYM50179

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9Br2Cl2NO2
CAS Number:
Molecular Weight:
441.93
UNSPSC Code:
12352211
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

CYM50179 is a potent, selective agonist of the sphingosine-1-phosphate receptor 4 (S1P4).
CYM50179 is a potent, selective agonist of the sphingosine-1-phosphate receptor 4 (S1P4). CYM50179 activates S1P4 with an IC50 of 46 nM, with no affect on other S1P receptors at concentrations up to 25 μM.

Features and Benefits

This compound is featured on the Lysophospholipid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Eye Dam. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sigma-Aldrich offers many products related to lysophospholipid receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service