Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0779

Sigma-Aldrich

RSVA314

≥98% (HPLC)

Synonym(s):

2-Hydroxy-benzoic acid 2-[[4-(diethylamino)-2-hydroxyphenyl]methylene]hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N3O3
CAS Number:
Molecular Weight:
327.38
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

light orange

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

RSVA314 is a resveratrol analog that is a potent indirect activator of AMPK (AMP-activated protein kinase).
RSVA314 is a resveratrol analog that is a potent indirect activator of AMPK (AMP-activated protein kinase). RSVA314 is a potent inhibitor of adipogenesis in vitro in cultured cells and in vivo in mice. Apparently RSVA314 activates AMPK via a mechanism implicating CaMKKβ and LKB1. Also RSVA314 increases intracellular clearance of the amyloidogenic peptide Aβ, the core component of the amyloid plaques.

Features and Benefits

This compound is featured on the AMPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The AMP-activated protein kinase (AMPK) acts as a sensor of cellular energy status. AMPK exists as heterotrimeric complexes comprising a catalytic α subunit and regulatory β and γ subunits.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service