Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0767

Sigma-Aldrich

NCGC00188636

≥98% (HPLC)

Synonym(s):

2-[(1,1-Dioxido-1,2-benzisothiazol-3-yl)thio]-benzoic acid, 4-[(1,1-Dioxo-1,2-benzothiazol-3-yl)sulfanyl]benzoic acid, DBS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9NO4S2
CAS Number:
Molecular Weight:
319.36
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

NCGC00188636 (DBS) is a potent and specific inhibitor of pyruvate kinase (PYK) that binds to the enzyme active site lysine residue (Lys 335 Leishmania mexicana PYK, Lys410 HsRPYK).
NCGC00188636 (DBS) is a potent and specific inhibitor of pyruvate kinase (PYK).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service