SML0758
WS3
≥98% (HPLC)
Synonym(s):
N-[6-[4-[[[[4-[(4-Methyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-amino]carbonyl]amino]phenoxy]-4-pyrimidinyl]-cyclopropanecarboxamide, WS-3
About This Item
Recommended Products
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 10 mg/mL, clear
storage temp.
2-8°C
InChI
1S/C28H30F3N7O3/c1-37-10-12-38(13-11-37)16-19-4-5-21(14-23(19)28(29,30)31)35-27(40)34-20-6-8-22(9-7-20)41-25-15-24(32-17-33-25)36-26(39)18-2-3-18/h4-9,14-15,17-18H,2-3,10-13,16H2,1H3,(H2,34,35,40)(H,32,33,36,39)
InChI key
KIKOYRNAERIVSJ-UHFFFAOYSA-N
Biochem/physiol Actions
Features and Benefits
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Our company offers many products related to EGFR for your research needs.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service