SML0726
Panepoxydone
from Lentinus conatus, ≥95% (HPLC)
Synonym(s):
(1S,5R,6S)-5-Hydroxy-3-[(1S)-1-hydroxy-3-methyl-2-buten-1-yl]-7-oxabicyclo[4.1.0]hept-3-en-2-one
About This Item
biological source
Lentinus conatus
assay
≥95% (HPLC)
form
powder
storage condition
protect from light
solubility
DMSO: soluble 10 mg/mL
Storage temp.
−20°C
InChI
1S/C11H14O4/c1-5(2)3-7(12)6-4-8(13)10-11(15-10)9(6)14/h3-4,7-8,10-13H,1-2H3/t7?,8-,10+,11-/m1/s1
Inchi Key
MBXKEYXHJAZKBP-VQZXEUHYSA-N
Biochem/physiol Actions
Reconstitution
Other Notes
signalword
Danger
hcodes
pcodes
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service