Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0721

Sigma-Aldrich

LSN2463359

≥98% (HPLC)

Synonym(s):

N-(1-Methylethyl)-5-(pridin-4-ylethynyl)pyridine-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N3O
CAS Number:
Molecular Weight:
265.31
UNSPSC Code:
12352202
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C16H15N3O/c1-12(2)19-16(20)15-6-5-14(11-18-15)4-3-13-7-9-17-10-8-13/h5-12H,1-2H3,(H,19,20)

InChI key

STAFRSGTRKNXHF-UHFFFAOYSA-N

Biochem/physiol Actions

LSN2463359 is a selactive brain penetrant mGluR5 positive allosteric modulator.
LSN2463359 is a selactive brain penetrant mGluR5 positive allosteric modulator. LSN2463359 is a potentiator of mGluR5-mediated Ca2+ influx in vitro, giving curve shifts 2 to 3 fold over agonist alone. In vivo, the compound exerts wake-promoting, cognitive and motor effects.

Features and Benefits

This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service