Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0718

Sigma-Aldrich

UBP714

≥95% (HPLC)

Synonym(s):

6-Bromo-4-methyl-2-oxo-2H-1-benzopyran-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7BrO4
CAS Number:
Molecular Weight:
283.07
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C11H7BrO4/c1-5-7-4-6(12)2-3-8(7)16-11(15)9(5)10(13)14/h2-4H,1H3,(H,13,14)

InChI key

BWBWVUJRXNIUMA-UHFFFAOYSA-N

Biochem/physiol Actions

UBP714 is a GluN1/GluN2-containing NMDA receptor positive allosteric modulator.
UBP714 is a derivative of the NMDA receptor negative allosteric modulator UBP608 (SML0606). UBP714 potentiates the activity of recombinant GluN1/GluN2 NMDA receptors, and enhances NMDA receptor mediated EPSPs in the CA1 region of the hippocampus.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service