Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0706

Sigma-Aldrich

2-AEMP trifluoroacetate salt

≥97% (NMR)

Synonym(s):

2-Aminoethyl methylphosphonate trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H10NO3P · CF3COOH
CAS Number:
Molecular Weight:
253.11
UNSPSC Code:
12352202
NACRES:
NA.77

assay

≥97% (NMR)

form

oil

color

colorless to light yellow

storage temp.

2-8°C

Biochem/physiol Actions

2-AEMP is a competitive GABAC antagonist with a rapid onset of, and release from inhibition.
2-AEMP is a competitive GABAC antagonist.

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Kammer et al.
Biosensors & bioelectronics, 131, 119-127 (2019-03-04)
Organophosphorus compounds (OPs) continue to represent a significant chemical threat to humans due to exposures from their use as weapons, their potential storage hazards, and from their continued use agriculturally. Existing methods for detection include ELISA and mass spectrometry. The

Articles

Introduction to GABAC receptors as well as modulators and other products for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service