Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0703

Sigma-Aldrich

MDPPP hydrochloride

≥98% (HPLC)

Synonym(s):

1-(1,3-Benzodioxol-5-yl)-2-(1-pyrrolidinyl)-1-propanone hydrochloride, 3′,4′-MDPPP, 3’,4’-MD-α-PPP hydrochloride, 3’,4’-Methylenedioxy-α-pyrrolidinopropiophenone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO3 · HCl
CAS Number:
Molecular Weight:
283.75
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

MDPPP hydrochloride is a potential designer drug structurally similar to cathinones, amphetamines, and pyrovalerone.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service