Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0677

Sigma-Aldrich

IWP12

≥98% (HPLC)

Synonym(s):

IWP-12, N-(6-Methyl-2-benzothiazolyl)-2-[(3,4,6,7-tetrahydro-3,6-dimethyl-4-oxothieno[3,2-d]pyrimidin-2-yl)thio]-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N4O2S3
CAS Number:
Molecular Weight:
418.56
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C18H18N4O2S3/c1-9-4-5-11-13(6-9)27-17(19-11)21-14(23)8-25-18-20-12-7-10(2)26-15(12)16(24)22(18)3/h4-6,10H,7-8H2,1-3H3,(H,19,21,23)

InChI key

RBFDSBJDWZOTGR-UHFFFAOYSA-N

Biochem/physiol Actions

IWP (inhibitor of Wnt production) inhibitors are known to prevent Wnt protein generation. Abnormal Wnt signalling associated with tumorigenesis and metastasis of tumors.[1]
IWP12 is Wnt inhibitor that prevents palmitylation of Wnt proteins by Porcupine (Porcn), which is suitable for in vivo assays. IWP12 blocked post resection juvenile zebrafish tailfin regeneration.
IWP12 is Wnt inhibitor.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The development of highly potent inhibitors for porcupine.
Wang X, et al.
Journal of Medicinal Chemistry, 56(6), 2700-2704 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service