Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0659

Sigma-Aldrich

LYG-202

≥98% (HPLC)

Synonym(s):

5-Hydroxy-8-methoxy-7-[4-(4-methyl-1-piperazinyl)butoxy]-2-phenyl-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30N2O5
CAS Number:
Molecular Weight:
438.52
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

LYG-202 is a synthetic flavanoid that induces apoptosis and is a CK2 inhibitor.
LYG-202 is a synthetic flavanoid that induces apoptosis in several cancer cell lines. LYG-202 induces G1/S phase cell cycle arrest and apoptosis in HCT116 cells by a mechanism that involves increases in p21(WAF1/Cip1) and Rb expression. In HepG2 cells, the compound LYG-202 potentiates the apoptotic effects of TNFα by inhibiting casein kinase 2 (CK2).

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faiza M Kalfalah et al.
Cell cycle (Georgetown, Tex.), 14(11), 1704-1715 (2015-04-16)
To maintain genome stability, the thousands of replication origins of mammalian genomes must only initiate replication once per cell cycle. This is achieved by a strict temporal separation of ongoing replication in S phase, and the formation of pre-replicative complexes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service