Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0658

Sigma-Aldrich

3-MFA

≥97% (HPLC)

Synonym(s):

2-(3-Methylphenyl)aminobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13NO2
CAS Number:
Molecular Weight:
227.26
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C14H13NO2/c1-10-5-4-6-11(9-10)15-13-8-3-2-7-12(13)14(16)17/h2-9,15H,1H3,(H,16,17)

InChI key

JODCQKTVDXZQEP-UHFFFAOYSA-N

Biochem/physiol Actions

3-MFA is a mefenamic acid analog that blocks the transient receptor potential melastatin 2 (TRPM2) channel.
3-MFA is a mefenamic acid analog that blocks the transient receptor potential melastatin 2 (TRPM2) channel. Unlike other TRPM2 blockers such as 2-APB and flufenamic acid, 2-APB does not induce release of calcium from intracellular stores.

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to transient receptor potential channels for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service