SML0655
Shiga toxin 1, B subunit, HIS-tagged
recombinant, expressed in E. coli, ≥95% (SDS-PAGE)
Synonym(s):
SLT1, STX1, STxB
About This Item
Recommended Products
recombinant
expressed in E. coli
Quality Level
assay
≥95% (SDS-PAGE)
form
lyophilized
suitability
endotoxin tested
shipped in
dry ice
storage temp.
−20°C
Biochem/physiol Actions
Stx1 is identical to Stx, while the Stx2 isoforms share less sequence similarity with Stx (∼60%) and are immunologically distinct. In spite of the differences in their amino acid sequence, all Stx isoforms share the same overall toxin structure and mechanism of action.
Shiga toxins consists of two polypeptides. An A chain and a B chain non-covalently associate with an apparent stoichiometry of one A and five B chains to form the holotoxin. The catalytic A subunit has
RNA N-glycosidase activity that inhibits eukaryotic protein synthesis. The B subunits form a pentamer, which recognizes and binds to the functional cell-surface receptor globotriaosylceramide [Gb3; Gala(1-4)-Galb(1-4)-Glcb1-ceramide]. Gb3 is overexpressed in membranes of numerous tumor cells, therefore STxB binding to Gb3 receptors may be useful for cell-specific vectorization, labeling, and imaging purposes.
Physical form
Preparation Note
Analysis Note
signalword
Warning
hcodes
pcodes
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service