Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0639

Sigma-Aldrich

Ro 11-1464

≥98% (HPLC)

Synonym(s):

9-Methyl-4-phenyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$390.00

$390.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
25 MG
$390.00

About This Item

Empirical Formula (Hill Notation):
C15H12N4S
CAS Number:
Molecular Weight:
280.35
UNSPSC Code:
12352202
NACRES:
NA.77

$390.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Ro 11-1464 induces the hepatic expression of apoA-1 and leads to increased levels of apoA-1 in the plasma. Treatment of human apoA-1 transgenic mice with Ro 11-1464 resulted in a three-fold increase in hapoA-1 mRNA in the liver, and a two-fold increase in plama apoA-1levels. The compound causes a significant decrease in plama lipids and enhances reverse cholesterol transport.
Ro 11-1464 induces the hepatic expression of apoA-1.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service