Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0637

Sigma-Aldrich

BD750

≥98% (HPLC)

Synonym(s):

2-(2-Benzothiazolyl)-4,5,6,7-tetrahydro-2H-indazol-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13N3OS
CAS Number:
Molecular Weight:
271.34
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

BD750 is a benzothiazole derivative that inhibits proliferation of T-cells by a mechanism that involves inhibition of STAT5 phosphorylation and expression of cyclin D3 and CDK6. In the presence of BD750, human and mouse T cells stimulated with anti-CD3/CD28, alloantigen, PMA or ConA undergo G0/G1 cell cycle arrest, but express CD25 and CD69 and secrete IL-2 and IL-4. BD750 inhibits delayed-type hypersensitivity in mice.
BD750 is a benzothiazole deriviative, JAK3/STAT5 pathway inhibitor; immunosuppressnt and T-cell proliferation inhibitor.

Features and Benefits

This compound is featured on the JAKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service