Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0631

Sigma-Aldrich

NE-100

≥98% (HPLC)

Synonym(s):

4-Methoxy-3-(2-phenylethoxy)-N,N-dipropyl-benzeneethanamine hydrochloride, N,N-Di-n-propyl-2-[4-methoxy-3-(2-phenylethoxy)phenyl] ethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H33NO2 · HCl
CAS Number:
Molecular Weight:
391.97
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 15 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C21H29NO2.ClH/c1-4-22(5-2)15-13-19-11-12-20(23-3)21(17-19)24-16-14-18-9-7-6-8-10-18;/h6-12,17H,4-5,13-16H2,1-3H3;1H

InChI key

LPMIMPZUXJWRJJ-UHFFFAOYSA-N

Biochem/physiol Actions

NE-100 is a potent and selective σ1 receptor antagonist with an IC50 value of 1.54 +/- 0.26 nM for σ1 receptors, > 55-fold selectivity over σ2 receptors and > 6000-fold selectivity over D1, D2, 5-HT1A, 5-HT2 and PCP receptors.
NE-100 is a potent and selective σ1 receptor antagonist.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamaki Ishima et al.
Progress in neuro-psychopharmacology & biological psychiatry, 32(7), 1656-1659 (2008-07-24)
In addition to acetylcholinesterase (AChE) inhibition, donepezil binds to sigma-1 receptors. In this study, we examined the effects of donepezil on nerve growth factor (NGF)-induced neurite outgrowth in PC12 cells. Donepezil significantly potentiated the NGF-induced neurite outgrowth in a concentration-dependent
José Manuel Entrena et al.
Pain, 143(3), 252-261 (2009-04-21)
We evaluated the role of sigma(1) receptors on capsaicin-induced mechanical hypersensitivity and on nociceptive pain induced by punctate mechanical stimuli, using wild-type and sigma(1) receptor knockout (sigma(1)-KO) mice and selective sigma(1) receptor-acting drugs. Mutation in sigma(1)-KO mice was confirmed by
Md Shenuarin Bhuiyan et al.
Expert opinion on therapeutic targets, 14(10), 1009-1022 (2010-08-21)
We here investigated the effect of sigma-1 receptor (Sig-1R) stimulation with fluvoxamine on myocardial hypertrophy, cardiac functional recovery and defined mechanisms underlying its cardioprotective action. Wistar rats subjected to bilateral ovariectomy (OVX) were treated with abdominal aortic banding between the
Takahito Yamamoto et al.
Current drug metabolism, 7(2), 135-146 (2006-02-14)
The purpose of this study was to propose a new method to predict in vivo drug-drug interactions (DDIs) for a high clearance drug from in vitro data. As the high clearance drug, NE-100 (N, N-dipropyl-2-[4-methoxy-3-(2-phenylethoxy)phenyl]ethylamine monohydrochloride) was used. First, approach
Yoko Hirata et al.
Journal of neurochemistry, 119(4), 839-847 (2011-09-03)
Chloroquine, a widely used anti-malarial and anti-rheumatoid agent, has been reported to induce apoptotic and non-apoptotic cell death. Accumulating evidence now suggests that chloroquine can sensitize cancer cells to cell death and augment chemotherapy-induced apoptosis by inhibiting autophagy. However, chloroquine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service