Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0626

Sigma-Aldrich

RFRP-3 human trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

NPVF, Neuropeptide VF (124-131), RFamide-related peptide-3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H72N14O10 · xC2HF3O2
CAS Number:
Molecular Weight:
969.14 (free base basis)
UNSPSC Code:
12352202
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C45H72N14O10/c1-24(2)21-31(57-40(65)30(23-35(47)61)56-42(67)33-15-10-20-59(33)44(69)36(48)25(3)4)43(68)58-19-9-14-32(58)41(66)54-28(16-17-34(46)60)39(64)53-27(13-8-18-52-45(50)51)38(63)55-29(37(49)62)22-26-11-6-5-7-12-26/h5-7,11-12,24-25,27-33,36H,8-10,13-23,48H2,1-4H3,(H2,46,60)(H2,47,61)(H2,49,62)(H,53,64)(H,54,66)(H,55,63)(H,56,67)(H,57,65)(H4,50,51,52)/t27-,28-,29-,30-,31-,32-,33-,36-/m0/s1

InChI key

JFZCLMZUABKABL-GVCDGELBSA-N

Amino Acid Sequence

Val-Pro-Asn-Leu-Pro-Gln-Arg-Phe-NH2

Biochem/physiol Actions

RFRP-3 (RFamide-related peptide-3) is a homolog of avian gonadotropin-inhibitory hormone. RFRP-3 binds to the GPR147 receptor (also known as neuropeptide FF receptor 1). RFRP-3 blocks release of gonadotropin from the pituitary, and inhibits FSH-, LH- and forskolin-stimulated cAMP accumulation and progesterone production in human granulosa-lutein cells.
RFRP-3 is a homolog of avian gonadotropin-inhibitory hormone. RFRP-3 binds to the GPR147 receptor (also known as neuropeptide FF receptor 1).

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service