Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0605

Sigma-Aldrich

IQ-1S

≥98% (HPLC)

Synonym(s):

11H-Indeno[1,2-b]quinoxalin-11-one oxime Sodium; IQ-1 sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H8N3O · Na
CAS Number:
Molecular Weight:
269.23
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

IQ-1S is an inhibitor of JNK kinases, with a preference for JNK3.
IQ-1S is an inhibitor of JNK kinases, with a preference for JNK3. Kd values for IQ-3 binding to JNK 1, 2 and 3 binding are 390, 360 and 87 nM, respectively. The compound inhibits JNK phosphorylation and NFkB /AP-1 transcriptional activity in human monocytic cells at low mM concentrations, and inhibits CD4+ T-cell immune response in a murine delayed-type hypersensitivity (DTH) model.

Features and Benefits

This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service