Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0600

Sigma-Aldrich

LPSF/GQ-02

≥98% (HPLC)

Synonym(s):

(5Z)-5-[(4-Chlorophenyl)methylene]-3-[(4-methylphenyl)methyl]-2,4-thiazolidinedione, 5-(4-Chloro-benzylidene)-3-(4-methyl-benzyl)-thiazolidine-2,4-dione, GQ-02

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14ClNO2S
CAS Number:
Molecular Weight:
343.83
UNSPSC Code:
12352202
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

LPSF/GQ-02 is a new thiazolidinedione that shows improved insulin resistance, reduces the area of atherosclerotic lesions, and offers a protective effect for the endothelium in LDL receptor-deficient mice.
LPSF/GQ-02 is a new thiazolidinedione that shows improved insulin resistance, reduces the area of atherosclerotic lesions, and offers a protective effect for the endothelium in LDL receptor-deficient mice. LPSF/GQ-02 induced an overexpression of eNOS and significantly inhibited the expression of metalloproteinases, both of which can result in anti-inflammatory effects.

Features and Benefits

This compound is featured on the Nuclear Receptors (PPARs) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service