Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0575

Sigma-Aldrich

BRD9539

≥98% (HPLC)

Synonym(s):

2-(Benzoylamino)-1-(3-phenylpropyl)-1H-benzimidazole-5-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21N3O3
CAS Number:
Molecular Weight:
399.44
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(=O)c1ccc2n(CCCc3ccccc3)c(NC(=O)c4ccccc4)nc2c1

InChI

1S/C24H21N3O3/c28-22(18-11-5-2-6-12-18)26-24-25-20-16-19(23(29)30)13-14-21(20)27(24)15-7-10-17-8-3-1-4-9-17/h1-6,8-9,11-14,16H,7,10,15H2,(H,29,30)(H,25,26,28)

InChI key

WPXMEOBILYVKBC-UHFFFAOYSA-N

Biochem/physiol Actions

BRD9539 is a very specific inhibitor of the enzymatic activity of histone methyltransferase G9a.
BRD9539 is a very specific inhibitor of the enzymatic activity of histone methyltransferase G9a. The compound has poor activity in cell based assays.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service