Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0554

Sigma-Aldrich

UNC926

≥98% (HPLC)

Synonym(s):

(3-Bromophenyl)[4-(1-pyrrolidinyl)-1-piperidinyl]-methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21BrN2O
CAS Number:
Molecular Weight:
337.25
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Brc1cccc(c1)C(=O)N2CCC(CC2)N3CCCC3

InChI

1S/C16H21BrN2O/c17-14-5-3-4-13(12-14)16(20)19-10-6-15(7-11-19)18-8-1-2-9-18/h3-5,12,15H,1-2,6-11H2

InChI key

OWGLFIKZKQOYHZ-UHFFFAOYSA-N

Biochem/physiol Actions

UNC926 is a specific antagonist of the methyl-lysine binding protein L3MBTL1, a member of a protein family that reads the methylation state of histone lysine residues to direct gene expression. UNC926 dose dependently blocks the interaction of recombinant L3MBTL carrying three tandem MBT reader domains (L3MBTL3XMBT) and a methylated histone peptide H4K20me1 with an IC50 of 3.9 μM.
UNC926 is a specific antagonist of the methyl-lysine binding protein L3MBTL1.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service