Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0532

Sigma-Aldrich

N-Benzyl-2,4-dinitrobenzenesulfonamide

≥98% (HPLC)

Synonym(s):

2,4-Dinitro-N-(phenylmethyl)-benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11N3O6S
CAS Number:
Molecular Weight:
337.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(c(c1)[N+]([O-])=O)S(=O)(=O)NCc2ccccc2

InChI

1S/C13H11N3O6S/c17-15(18)11-6-7-13(12(8-11)16(19)20)23(21,22)14-9-10-4-2-1-3-5-10/h1-8,14H,9H2

InChI key

XKHQYTSLQZPZOF-UHFFFAOYSA-N

Biochem/physiol Actions

N-benzyl-2,4-dinitrobenzenesulfonamide is a thiol activated source of SO2.
N-benzyl-2,4-dinitrobenzenesulfonamide reacts with cysteine to release SO2.

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service