Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0520

Sigma-Aldrich

Arvanil

≥98% (HPLC), solution (in ethanol)

Synonym(s):

(5Z,8Z,11Z,14Z)-N-[(4-Hydroxy-3-methoxyphenyl)methyl]-5,8,11,14-Eicosatetraenamide, N-Vanillylarachidonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H41NO3
CAS Number:
Molecular Weight:
439.63
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

form

solution (in ethanol)

concentration

50 mg/mL in ethanol

color

colorless to pale yellow

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)NCc1ccc(O)c(OC)c1

InChI

1S/C28H41NO3/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-28(31)29-24-25-21-22-26(30)27(23-25)32-2/h7-8,10-11,13-14,16-17,21-23,30H,3-6,9,12,15,18-20,24H2,1-2H3,(H,29,31)/b8-7-,11-10-,14-13-,17-16-

InChI key

QVLMCRFQGHWOPM-ZKWNWVNESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Arvanil is an activator of cannabinoid and vanilloid receptors, and a potent inhibitor of anandamide accumulation. Arvanil inhibits growth of astrocytoma xenograft tumors in mice, mimicing the effect of neural precursor cells, that have been shown to kill TRPV1-expressing tumor cells by secreting endovanilloids.
Arvanil is an.activator of cannabinoid and vanilloid receptors; potent inhibitor of anandamide accumulation.

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Light and air sensitive

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

57.2 °F - closed cup

flash_point_c

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen D Richbart et al.
Journal of medicinal chemistry, 64(3), 1346-1361 (2021-01-29)
Capsaicin displays robust growth-inhibitory activity in multiple human cancers. However, the feasibility of capsaicin as a clinically relevant anticancer drug is hampered by its adverse side effects. This concern has led to extensive research focused on the isolation and synthesis
Sofia Sisay et al.
PloS one, 8(10), e76907-e76907 (2013-10-17)
Endocannabinoids and some phytocannabinoids bind to CB1 and CB2 cannabinoid receptors, transient receptor potential vanilloid one (TRPV1) receptor and the orphan G protein receptor fifty-five (GPR55). Studies using C57BL/10 and C57BL/6 (Cnr2 (tm1Zim)) CB2 cannabinoid receptor knockout mice have demonstrated
Anna Zaia Carolina Rodrigues et al.
Molecular and cellular neurosciences, 95, 59-70 (2019-02-15)
Increasing evidence indicates that, first, the sympathetic nervous system interacts extensively with both vasculature and skeletal muscle fibers near neuromuscular junctions (NMJs) and, second, its neurotransmitter, noradrenaline, influences myofiber molecular composition and function and motor innervation. Since sympathomimetic agents have

Articles

We offer many products related to cannabinoid receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service