Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0514

Sigma-Aldrich

UCM710

≥98% (HPLC)

Synonym(s):

(9Z)-9-Hexadecenoic acid 2-oxiranylmethyl ester, (±)-Oxiran-2-ylmethyl (9Z)-hexadec-9-enoate

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$353.00

$353.00


Available to ship onApril 28, 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
$353.00

About This Item

Empirical Formula (Hill Notation):
C19H34O3
CAS Number:
Molecular Weight:
310.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$353.00


Available to ship onApril 28, 2025Details


Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

oil

color

colorless to light yellow

storage temp.

2-8°C

SMILES string

CCCCCC\C=C/CCCCCCCC(=O)OCC1CO1

InChI

1S/C19H34O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(20)22-17-18-16-21-18/h7-8,18H,2-6,9-17H2,1H3/b8-7-

InChI key

FWEHVPCBXKCYHE-FPLPWBNLSA-N

Biochem/physiol Actions

UCM710 is a dual inhibitor of FAAH and ABHD6.
UCM710 is a dual inhibitor of fatty acid amide hydrolase (FAAH) and α/β hydrolase domain 6 (ABHD6) the enzymes that hydrolyze endocannabinoids. UCM710 augments both N-arachidonoylethanolamine (AEA) and 2-arachidonoylglycerol (2-AG) levels in neurons. UCM710 does not inhibit monoacylglycerol lipase (MAGL).

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to cannabinoid receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service