Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0498

Sigma-Aldrich

PNU112455A

≥98% (HPLC)

Synonym(s):

4-[(6-amino-4-pyrimidinyl)amino]-benzenesulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N5O2S · HCl
CAS Number:
Molecular Weight:
301.75
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

Cl.Nc1cc(Nc2ccc(cc2)S(N)(=O)=O)ncn1

InChI

1S/C10H11N5O2S.ClH/c11-9-5-10(14-6-13-9)15-7-1-3-8(4-2-7)18(12,16)17;/h1-6H,(H2,12,16,17)(H3,11,13,14,15);1H

InChI key

XBXQRCJKDCEQBS-UHFFFAOYSA-N

Biochem/physiol Actions

PNU112455A is a CDK 2/5 inhibitor.
PNU112455A is an ATP site competetive inhibitor of CDK2 and CDK5. The Km for binding to the ATP site of CDK2 and CDK5 is 3.6 and 3.3 mM, respectively.

Features and Benefits

This compound is featured on the CDKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sholeem Griffin et al.
Biology of reproduction, 99(4), 749-760 (2018-04-25)
Preventing postpartum uterine disease depends on the ability of endometrial cells to tolerate the presence of the bacteria that invade the uterus after parturition. Postpartum uterine disease and endometrial pathology in cattle are most associated with the pathogen Trueperella pyogenes.

Articles

Review properties, activators and inhibitors, and available products for researching cyclin-dependent kinases (CDKs).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service