Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0457

Sigma-Aldrich

AE9C90CB

≥98% (HPLC)

Synonym(s):

N-[(1a,5a,6a)-3-Azabicyclo[3.1.0]hex-6-ylmethyl]-a-hydroxy-N-methyl-a-phenyl-benzeneacetamide hydrochloride (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24N2O2 · HCl
CAS Number:
Molecular Weight:
372.89
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 15 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

Cl.CN(C[C@@H]1[C@H]2CNC[C@@H]12)C(=O)C(O)(c3ccccc3)c4ccccc4

InChI

1S/C21H24N2O2.ClH/c1-23(14-19-17-12-22-13-18(17)19)20(24)21(25,15-8-4-2-5-9-15)16-10-6-3-7-11-16;/h2-11,17-19,22,25H,12-14H2,1H3;1H/t17-,18+,19+;

InChI key

YHBAWCACDJQGDL-MSFHQNQXSA-N

Biochem/physiol Actions

AE9C90CB is a muscarinic acetylcholine receptor antagonist with 20-fold selectivity for M3 receptor subtypes compared to M2. In an in vivo assay, AE9C90CB was approximately 5 times more selective for inhibiting carbachol-induced urinary bladder contraction, compared to saliva production.
AE9C90CB is a tissue specific M3 muscarinic receptor antagonist.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Muscarinic acetylcholine receptors mediate acetylcholine actions in CNS and non-nervous tissues, crucial for cell signaling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service