Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0426

Sigma-Aldrich

NPD4456

≥98% (HPLC)

Synonym(s):

Dimethyl-carbamic acid 3-(2,5-dimethoxyphenyl)-4-methyl-2-oxo-2H-1-benzopyran-7-yl ester (9CI), RIKEN Natural Product Derivative (NPD) 4456

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21NO6
CAS Number:
Molecular Weight:
383.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (warmed; clear solution)

storage temp.

2-8°C

SMILES string

COc1ccc(OC)c(c1)C2=C(C)c3ccc(OC(=O)N(C)C)cc3OC2=O

InChI

1S/C21H21NO6/c1-12-15-8-6-14(27-21(24)22(2)3)11-18(15)28-20(23)19(12)16-10-13(25-4)7-9-17(16)26-5/h6-11H,1-5H3

InChI key

NHTDUXSQDZDSKQ-UHFFFAOYSA-N

Biochem/physiol Actions

NPD4456 is an HIV-1 Vpr inhibitor that directly binds to HIV-1 Vpr. NPD4456 inhibits Vpr-dependent viral infection of human macrophages. Vipirinin analogue.
NPD4456 is an HIV-1 Vpr inhibitor.

pictograms

Environment

signalword

Warning

hcodes

pcodes

Hazard Classifications

Aquatic Acute 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service