Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0401

Sigma-Aldrich

RBx-0597

≥96% (HPLC)

Synonym(s):

(2S,4S)-4-Fluoro-1-[2-[[(1a, 5a, 6a)-3-(4-fluorobenzoyl)-3-azabicyclo[3.1.0]hex-6-yl]amino]acetyl]-2-pyrrolidinecarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20F2N4O2
CAS Number:
Molecular Weight:
374.38
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

assay

≥96% (HPLC)

form

film

storage condition

desiccated

color

colorless

solubility

H2O: 2 mg/mL (clear solution)

storage temp.

−20°C

SMILES string

F[C@H]1C[C@@H](C#N)N(C1)C(=O)CN[C@@H]2[C@H]3CN(C[C@@H]23)C(=O)c4ccc(F)cc4

InChI

1S/C19H20F2N4O2/c20-12-3-1-11(2-4-12)19(27)24-9-15-16(10-24)18(15)23-7-17(26)25-8-13(21)5-14(25)6-22/h1-4,13-16,18,23H,5,7-10H2/t13-,14-,15-,16+,18+/m0/s1

InChI key

RWFPPHFEFUKAQS-YXISZKLHSA-N

Biochem/physiol Actions

RBx-0597 is a potent, competitive inhibitor of DPP-IV (IC50 = 30 nM), that displays selectivity or 150 and 300 fold over DDP-VIII and DDP-IX, respectively.
RBx-0597 is a potent, competitive inhibitor of DPP-IV.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service