Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0386

Sigma-Aldrich

CUR61414

≥98% (HPLC)

Synonym(s):

N-[(3S,5S)-1-(1,3-Benzodioxol-5-ylmethyl)-5-(1-piperazinylcarbonyl)-3-pyrrolidinyl]-N-[(3-methoxyphenyl)methyl]-3,3-dimethyl-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H42N4O5
CAS Number:
Molecular Weight:
550.69
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL (clear solution, warmed)

storage temp.

−20°C

SMILES string

COc1cccc(CN([C@H]2C[C@@H](N(C2)Cc3ccc4OCOc4c3)C(=O)N5CCNCC5)C(=O)CC(C)(C)C)c1

InChI

1S/C31H42N4O5/c1-31(2,3)17-29(36)35(19-22-6-5-7-25(14-22)38-4)24-16-26(30(37)33-12-10-32-11-13-33)34(20-24)18-23-8-9-27-28(15-23)40-21-39-27/h5-9,14-15,24,26,32H,10-13,16-21H2,1-4H3/t24-,26+/m0/s1

InChI key

WPHXYKUPFJRJDK-AZGAKELHSA-N

Biochem/physiol Actions

CUR61414 is a Hedgehog pathway inhibitor; Smo antagonist.
CUR61414 specifically binds to, and antagonizes the activity of smoothened (Smo) in the hedgehog signaling pathway. CUR61414 inhibits proliferation and induces apoptosis in tumor cells without affecting non-tumor cells.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service