Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0383

Sigma-Aldrich

KM91104

≥98% (HPLC)

Synonym(s):

3,4-Dihydroxy-N’-(2-hydroxybenzylidene)benzohydrazide, 3,4-Dihydroxy-benzoic acid [(2-hydroxyphenyl)methylene]hydrazide, KM 91104

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N2O4
CAS Number:
Molecular Weight:
272.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >15 mg/mL

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1O)C(=O)N\N=C\c2ccccc2O

InChI

1S/C14H12N2O4/c17-11-4-2-1-3-10(11)8-15-16-14(20)9-5-6-12(18)13(19)7-9/h1-8,17-19H,(H,16,20)/b15-8+

InChI key

GWVYHPUGEQGQSF-OVCLIPMQSA-N

Biochem/physiol Actions

KM91104 is a cell-permeable vacuolar ATPases (V-ATPase) inhibitor that specifically targets the V-ATPase a3-B2 subunits interaction. KM91104 inhibits osteoclast resorption.
KM91104 is a cell-permeable vacuolar ATPases (V-ATPase) inhibitor.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service