Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0380

Sigma-Aldrich

FMP-API-1

≥98% (HPLC)

Synonym(s):

3,3′-Diamino-4,4′-dihydroxydiphenylmethane, 4,4′-Methylenebis(2-aminophenol), Bis(3-amino-4-hydroxyphenyl)methane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14N2O2
CAS Number:
Molecular Weight:
230.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

SMILES string

Nc1cc(Cc2ccc(O)c(N)c2)ccc1O

InChI

1S/C13H14N2O2/c14-10-6-8(1-3-12(10)16)5-9-2-4-13(17)11(15)7-9/h1-4,6-7,16-17H,5,14-15H2

InChI key

KCFVSHSJPIVGCG-UHFFFAOYSA-N

Biochem/physiol Actions

FMP-API-1 is an inhibitor of AKAP-PKA interactions in vitro and in cultured cardiac myocytes that activates PKA. Apparently, FMP-API-1 binds to a novel allosteric regulatory site.
FMP-API-1 is an inhibitor of AKAP-PKA interactions.

Features and Benefits

This compound is featured on the PKA & PKG page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Air Sensitive

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fumiaki Ando et al.
Nature communications, 9(1), 1411-1411 (2018-04-14)
Congenital nephrogenic diabetes insipidus (NDI) is characterized by the inability of the kidney to concentrate urine. Congenital NDI is mainly caused by loss-of-function mutations in the vasopressin type 2 receptor (V2R), leading to impaired aquaporin-2 (AQP2) water channel activity. So
Louis-Philippe Bernier et al.
Cell reports, 27(10), 2895-2908 (2019-06-06)
Microglia, the brain's immune cells, maintain homeostasis and sense pathological changes by continuously surveying the parenchyma with highly motile large processes. Here, we demonstrate that microglia also use thin actin-dependent filopodia that allow fast nanoscale sensing within discrete regions. Filopodia

Articles

Our offers many products related to PKA & PKG for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service