Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0368

Sigma-Aldrich

CHD-5

≥98% (HPLC)

Synonym(s):

N-[2-Methyl-4-[2-(2-methylphenyl)diazenyl]phenyl]-2-furancarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N3O2
CAS Number:
Molecular Weight:
319.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

light orange to dark orange

solubility

DMSO: >15 mg/mL

storage temp.

2-8°C

SMILES string

Cc1ccccc1\N=N\c2ccc(NC(=O)c3ccco3)c(C)c2

InChI

1S/C19H17N3O2/c1-13-6-3-4-7-17(13)22-21-15-9-10-16(14(2)12-15)20-19(23)18-8-5-11-24-18/h3-12H,1-2H3,(H,20,23)/b22-21+

InChI key

RDQWQMVBBQKHGH-QURGRASLSA-N

Biochem/physiol Actions

CHD-5 is a potent aryl hydrocarbon receptor antagonist.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to non-steroid nuclear receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service