Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0316

Sigma-Aldrich

HZ52

≥98% (HPLC)

Synonym(s):

2-[[4-([1,1′-Biphenyl]-4-ylamino)-6-chloro-2-pyrimidinyl]thio]-octanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26ClN3O2S
CAS Number:
Molecular Weight:
456.00
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

ClC1=NC(SC(CCCCCC)C(O)=O)=NC(NC(C=C2)=CC=C2C3=CC=CC=C3)=C1

InChI

1S/C24H26ClN3O2S/c1-2-3-4-8-11-20(23(29)30)31-24-27-21(25)16-22(28-24)26-19-14-12-18(13-15-19)17-9-6-5-7-10-17/h5-7,9-10,12-16,20H,2-4,8,11H2,1H3,(H,29,30)(H,26,27,28)

InChI key

MLNIGXYHGQLWLD-UHFFFAOYSA-N

Biochem/physiol Actions

HZ52 is a dual inhibitor of mPGES and 5-lypoxygenase (Enzyme IC50 = 1.3 and 2.0 mM, respectively). HZ52 inhibits carrageenan-induced pleurisy and PAF-induced lethal shock in mouse.
HZ52 is a dual inhibitor of mPGES and 5-lypoxygenase.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eleni C Georgiadi et al.
Leukemia & lymphoma, 56(3), 748-754 (2014-06-14)
E2F-1 is the best-described member of the E2F family of transcriptional factors and is particularly interesting in view of its often opposing roles. Our purpose was to examine the immunohistochemical expression of E2F-1 in Hodgkin lymphoma (HL) and to correlate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service