Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0267

Sigma-Aldrich

NSC 162535 disodium salt

≥98% (HPLC)

Synonym(s):

7-Amino-4-hydroxy-3-[2-(5-hydroxy-7-sulfo-2-naphthalenyl)diazenyl]-2-naphthalenesulfonic acid sodium salt (1:2), Disodium 7-amino-4-hydroxy-3-[(5-hydroxy-7-sulfonatonaphthalen-2-yl)diazenyl]naphthalene-2-sulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13N3O8S2 · 2Na
CAS Number:
Molecular Weight:
533.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

faintly red to very dark red

solubility

H2O: ≥2 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

[Na+].[Na+].Nc1ccc2c(O)c(\N=N\c3ccc4c(O)cc(cc4c3)S([O-])(=O)=O)c(cc2c1)S([O-])(=O)=O

InChI

1S/C20H15N3O8S2.2Na/c21-12-1-3-16-10(5-12)8-18(33(29,30)31)19(20(16)25)23-22-13-2-4-15-11(6-13)7-14(9-17(15)24)32(26,27)28;;/h1-9,24-25H,21H2,(H,26,27,28)(H,29,30,31);;/q;2*+1/p-2/b23-22+;;

InChI key

ISVGEQZXHOLKRA-VPMNAVQSSA-L

Biochem/physiol Actions

NSC 162535 is an inhibitor of RNA editing in trypanosomatid pathogens that results from lost of the interaction of core editosome with substrate RNA.
NSC 162535 is an inhibitor of RNA editing in trypanosomatid pathogens.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service