Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0250

Sigma-Aldrich

anti-3-OS HS peptide trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

G2 trifluoroacetate salt, L-Methionyl-L-prolyl-L-arginyl-L-arginyl-L-arginyl-L-arginyl-L-isoleucyl-L-arginyl-L-arginyl-L-arginyl-L-glutaminyl-L-lysine trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C69H133N35O14S · xC2HF3O2
CAS Number:
Molecular Weight:
1709.09 (free base basis)
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

lyophilized powder

color

white

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC[C@H](C)[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H]1CCCN1C(=O)[C@@H](N)CCSC)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCCCN)C(O)=O

InChI

1S/C69H133N35O14S.C2HF3O2/c1-4-37(2)50(60(115)101-44(21-12-33-92-68(83)84)56(111)97-40(17-8-29-88-64(75)76)52(107)95-42(19-10-31-90-66(79)80)54(109)99-46(24-25-49(72)105)57(112)102-47(62(117)118)15-5-6-27-70)103-58(113)45(22-13-34-93-69(85)86)98-53(108)41(18-9-30-89-65(77)78)94-51(106)39(16-7-28-87-63(73)74)96-55(110)43(20-11-32-91-67(81)82)100-59(114)48-23-14-35-104(48)61(116)38(71)26-36-119-3;3-2(4,5)1(6)7/h37-48,50H,4-36,70-71H2,1-3H3,(H2,72,105)(H,94,106)(H,95,107)(H,96,110)(H,97,111)(H,98,108)(H,99,109)(H,100,114)(H,101,115)(H,102,112)(H,103,113)(H,117,118)(H4,73,74,87)(H4,75,76,88)(H4,77,78,89)(H4,79,80,90)(H4,81,82,91)(H4,83,84,92)(H4,85,86,93);(H,6,7)/t37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-,50-;/m0./s1

InChI key

FSDZLKPARBNKET-UTYCFSRRSA-N

Amino Acid Sequence

Met-Pro-Arg-Arg-Arg-Arg-Ile-Arg-Arg-Arg-Gln-Lys trifluoroacetate salt

Biochem/physiol Actions

MPRRRRIRRRQK is an anti-3-OS HS peptide that binds 3-O sulfated heparan sulfate (3-OS HS) and prevents HSV-1 infection in vivo and in cell culture. It also prevents entry of other members of herpesviruses family including cytomegalovirus (CMV) and human herpesvirus-8 (HHV-8). MPRRRRIRRRQK exhibits protective effects against HSV-1 infection of the mouse cornea.
anti-3-OS HS peptide is a 3-O sulfated heparan sulfate (3-OS HS) ligand that prevent HSV-1 infection in vivo and in cell culture.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service