Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0236

Sigma-Aldrich

ZM 336372

≥98% (HPLC)

Synonym(s):

3-(Dimethylamino)-N-[3-[(4-hydroxybenzoyl)amino]-4-methylphenyl]-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H23N3O3
CAS Number:
Molecular Weight:
389.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >5 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

CN(C)c1cccc(c1)C(=O)Nc2ccc(C)c(NC(=O)c3ccc(O)cc3)c2

InChI

1S/C23H23N3O3/c1-15-7-10-18(24-23(29)17-5-4-6-19(13-17)26(2)3)14-21(15)25-22(28)16-8-11-20(27)12-9-16/h4-14,27H,1-3H3,(H,24,29)(H,25,28)

InChI key

PYEFPDQFAZNXLI-UHFFFAOYSA-N

Biochem/physiol Actions

ZM-336372 is a potent inhibitor of the MAP kinase c-raf in in vitro assay systems (IC50 = 70 nM). In cell-based experiments, ZM-336372 induces c-raf activation through a novel feedback loop in which inhibition is counterbalanced by reactivation.
ZM-336372 is a potent inhibitor of the MAP kinase c-raf.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 4 - Skin Sens. 1B

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Weissinger et al.
British journal of cancer, 91(1), 186-192 (2004-06-10)
Activation of PKA by cAMP agonists, such as 8-Cl-cAMP activation, selectively causes rapid apoptosis in v-abl transformed fibroblasts by inhibiting the Raf-1 kinase. Here we investigated whether 8-Cl-cAMP is useful for the treatment of chronic myelogenous leukaemia (CML), which is

Articles

The extracellular signal regulated kinase (ERK1 and ERK2) pathways are activated by mitogens and play an important role in controlling cell growth and differentiation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service