Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0194

Sigma-Aldrich

3,4-Methylenedioxypyrovalerone hydrochloride

≥98% (HPLC)

Synonym(s):

1-(1,3-Benzodioxol-5-yl)-2-(1-pyrrolidinyl)-1-pentanone hydrochloride, MDPV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21NO3 · HCl
CAS Number:
Molecular Weight:
311.80
UNSPSC Code:
12352200

assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule I; Home Office Schedule 1; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

color

white to tan

solubility

H2O: ≥10 mg/mL

storage temp.

2-8°C

InChI

1S/C16H21NO3.ClH/c1-2-5-13(17-8-3-4-9-17)16(18)12-6-7-14-15(10-12)20-11-19-14;/h6-7,10,13H,2-5,8-9,11H2,1H3;1H

InChI key

PYQZNWFQAMFAMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,4-Methylenedioxypyrovalerone hydrochloride may be used in dopamine-mediated cell signaling studies.

Biochem/physiol Actions

3,4-Methylenedioxypyrovalerone is a derivative of pyrovalerone, a rapidly-absorbed psychostimulant. It belongs to the class of synthetic cathinones often used as performance-enhancing agent by athletes before being banned.[1][2]
3,4-Methylenedioxypyrovalerone is a norepinephrine-dopamine reuptake inhibitor (NDRI) that acts as a CNS stimulant.
3,4-Methylenedioxypyrovalerone is a norepinephrine-dopamine reuptake inhibitor (NDRI); CNS stimulant.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Coppola et al.
Toxicology letters, 208(1), 12-15 (2011-10-20)
The illicit marketplace of substances of abuse continually offers for sale legal alternatives to controlled drugs to a large public. In recent years, a new group of designer drugs, the synthetic cathinones, has emerged as a new trend, particularly among
The metabolism of pyrovalerone hydrochloride.
W Michaelis et al.
Journal of medicinal chemistry, 13(3), 497-503 (1970-05-01)

Articles

We offer many products related to dopamine receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service