SML0193
JFD01307SC
≥95% (HPLC)
Synonym(s):
N-(Tetrahydro-1,1-dioxido-3-thienyl)-glycine
About This Item
Recommended Products
assay
≥95% (HPLC)
form
powder
storage condition
desiccated
color
white to tan
solubility
DMSO: ≥5 mg/mL at ~60 °C
storage temp.
2-8°C
SMILES string
OCC(=O)CNC1CCS(=O)(=O)C1
InChI
1S/C7H13NO4S/c9-4-7(10)3-8-6-1-2-13(11,12)5-6/h6,8-9H,1-5H2
InChI key
JQQXDZXXSDLRBC-UHFFFAOYSA-N
Application
Biochem/physiol Actions
Features and Benefits
Storage Class
13 - Non Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Glutamine's role in neurotransmitter synthesis and transport highlights its importance in neuronal function and glutamate production.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service