Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0182

Sigma-Aldrich

bPiDl hydrate

≥98% (HPLC)

Synonym(s):

1,1′-(1,10-decanediyl)bis[3-methyl-pyridinium iodide (1:2) hydrate, N,N-decane-1,10-diyl-bis-3-picolinium diiodide hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$165.00

$165.00


Available to ship onApril 29, 2025Details



Select a Size

Change View
10 MG
$165.00

About This Item

Empirical Formula (Hill Notation):
C22H34I2N2 · xH2O
CAS Number:
Molecular Weight:
580.33 (anhydrous basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$165.00


Available to ship onApril 29, 2025Details


Quality Level

assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

H2O: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

O.[I-].[I-].Cc1ccc[n+](CCCCCCCCCC[n+]2cccc(C)c2)c1

InChI

1S/C22H34N2.2HI.H2O/c1-21-13-11-17-23(19-21)15-9-7-5-3-4-6-8-10-16-24-18-12-14-22(2)20-24;;;/h11-14,17-20H,3-10,15-16H2,1-2H3;2*1H;1H2/q+2;;;/p-2

InChI key

ILAREMIWZYIULF-UHFFFAOYSA-L

Application

bPiDl hydrate may be used to study nicotinic receptor-mediated signaling.

Biochem/physiol Actions

bPiDI is an α6β2-specific antagonist that inhibits nicotine-evoked and endogenous dopamine release from rat striatal slices (IC50 = 150 nM, Imax = 55%). bPiDI treatment decreases nicotine self-administration, and nicotine-induced locomotor activity in rats.
bPiDl is a selective α6β2-specific nicotinic receptor antagonist

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sigma-Aldrich offers many products related to nicotinic acetylcholine receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service