Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0166

Sigma-Aldrich

Histamine H1 receptor Antagonist (H1-BY633-AN) synthetic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H60N7O6F2BS
Molecular Weight:
947.94
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥97% (HPLC)

storage temp.

−20°C

Biochem/physiol Actions

The SML0166 ligand was shown to antagonize the activity of the H1 agonist, histamine, in the ChanTest recombinant CHO cell line expressing the human H1 receptor, co-transfected with a serum response element-induced secreted placental alkaline phosphatase (SPAP) reporter gene.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Reconstitution

Dissolve 0.2 mg in 21.1 ul of DMSO to give a 10mM stock solution. Once reconstituted into DMSO the product, must be stored, preferably in smaller aliquots, at -20°C.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to histamine receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service