Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0163

Sigma-Aldrich

Beta-3 Adrenoceptor Partial Agonist (β3-633-AG) synthetic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H55BF2N6O7S
Molecular Weight:
932.88
UNSPSC Code:
51111800
NACRES:
NA.77

assay

≥97% (HPLC)

Quality Level

storage temp.

−20°C

Biochem/physiol Actions

This ligand is a partial agonist at β3 adrenoceptors, is a very weak partial agonist at β1, and has no agonist activity at β2. It also acts as an antagonist at β1, β2 and β3.

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Reconstitution

Dissolve 0.2 mg in 21.4 ul of DMSO to give a 10mM stock solution. Once reconstituted into DMSO the product, must be stored, preferably in smaller aliquots, at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

β-Adrenoceptors are activated either by norepinephrine or epinephrine. Learn more about the role of β-adrenoceptors in protein expression, cell signaling, and metabolism research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service