SML0162
Fluorescent Beta-2 Adrenoceptor Antagonist (β3-633-AN4) synthetic
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C46H53BF2N6O7S
Molecular Weight:
882.82
UNSPSC Code:
12352200
assay
≥97% (HPLC)
storage temp.
−20°C
Biochem/physiol Actions
The SML0162 ligand is an antagonist at β2, β1 and β3 adrenoceptors (in rank order of affinity).
Features and Benefits
This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
Reconstitution
Dissolve 0.2 mg in 22.7 ul of DMSO to give a 10mM stock solution. Once reconstituted into DMSO the product, must be stored, preferably in smaller aliquots, at -20°C.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service