Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0126

Sigma-Aldrich

OBA-09 hydrate

≥98% (HPLC)

Synonym(s):

2-(1,2-Dioxopropoxy)benzoic acid hydrate, O-Pyruvoylsalicylic acid hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O5 · xH2O
CAS Number:
Molecular Weight:
208.17 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥24 mg/mL

storage temp.

2-8°C

SMILES string

O.CC(=O)C(=O)Oc1ccccc1C(O)=O

InChI

1S/C10H8O5.H2O/c1-6(11)10(14)15-8-5-3-2-4-7(8)9(12)13;/h2-5H,1H3,(H,12,13);1H2

InChI key

DLKFQAHWYMEWIK-UHFFFAOYSA-N

Application

OBA-09 hydrate may be used in cell signaling studies.

Biochem/physiol Actions

Neuroprotective agent
OBA-09 is a neuroprotective agent. It is an ester of pyruvate and salicylic acid that acts as a multi-functional agent to protect against ischemic damages to the brain. Administration of 5 mpk OBA-09 following middle cerebral artery occlusion (MCAO) or oxygen-glucose deprivation (OGD) models of ischemic injury in rats prevents lipid peroxidation, ROS production and causes the recovery of NAD and ATP concentrations to near basal levels. The compound leads to reduction of infarct volume and also accelerates the recovery of behavioral and neurological deficits.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service