SML0110
Ilicicolin F
≥98% (HPLC), from Verticillium hemipterigenum
Synonym(s):
8′-Acetoxyascochlorin, Acetoxy-ascochlorin, Antibiotic LL-Z 1272 ζ, Benzaldehyde, 3-[(2E,4E)-5-[(1S,2S,3S,6R)-3-(acetyloxy)-1,2,6-trimethyl-5-oxocyclohexyl]-3-methyl-2,4-pentadien-1-yl]-5-chloro-2,4-dihydroxy-6-methyl
About This Item
biological source
Verticillium hemipterigenum
assay
≥98% (HPLC)
form
solid
solubility
DMSO: ≥10 mg/mL
H2O: insoluble
chloroform: soluble
methanol: soluble
storage temp.
−20°C
Application
Biochem/physiol Actions
Compounds of the ascochlorin family such as Ilicicolin F, C, D and H show a wide range of inhibitory effects on farnesyl protein transferase (FPTase) activity, and a significant inhibitory effect on the activity of testosterone-5α-reductase. Ilicicolin C and F have a moderate inhibitory activity toward the enzymes acetylcholinesterase (AChE) and β-glucuronidase and are active against Pseudomonas syringae with IC50 values of 28.5 μg/mL.
All isolated ascochlorin analogs exhibit significant antitumor and cytotoxic activities. Ascochlorin and its homologues are usable in treating and/or preventing diseases that can be relieved by the retinoid X receptor ligand-dependent signal transcriptional regulation (i.e. hypertension, cerebrovascular diseases, rheumatoid arthritis, autoimmune diseases, complication of diabetes, arteriosclerosis etc.). Moreover, they can inhibit denaturation and/or necrosis of pancreatic Langerhans islet β-cells and therefore can sustain insulin productivity.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service