Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0109

Sigma-Aldrich

GLYX-13 trifluoroacetate

≥98% (HPLC)

Synonym(s):

L-Threonyl-L-prolyl-L-prolyl-L-threoninamide trifluoroacetate; Thr-Pro-Pro-Thr-NH2 trifluoroacetate, Rapastinel, TPPT-amide trifluoroacetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$220.00

$220.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 MG
$220.00

About This Item

Empirical Formula (Hill Notation):
C18H31N5O6 · xC2HF3O2
CAS Number:
Molecular Weight:
413.47 (free base basis)
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

$220.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

≥98% (HPLC)

form

powder

color

white to tan

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.C[C@@H](O)[C@H](N)C(=O)N1CCC[C@H]1C(=O)N2CCC[C@H]2C(=O)N[C@@H]([C@@H](C)O)C(N)=O

InChI

1S/C18H31N5O6.C2HF3O2/c1-9(24)13(19)18(29)23-8-4-6-12(23)17(28)22-7-3-5-11(22)16(27)21-14(10(2)25)15(20)26;3-2(4,5)1(6)7/h9-14,24-25H,3-8,19H2,1-2H3,(H2,20,26)(H,21,27);(H,6,7)/t9-,10-,11+,12+,13+,14+;/m1./s1

InChI key

BCWVNVCGYWOAAK-GDLIIDCZSA-N

Application

GLYX-13 trifluoroacetate has been used as an activator of NMDA (N-methyl-D-aspartate) receptor.[1]

Biochem/physiol Actions

GLYX-13 is a glycine-site functional partial agonist (GFPA) selective modulator of the NMDA receptor (NMDAR). It is a brain penetrable tetrapeptide which acts as a positive allosteric modulator of the NMDA receptor, acting via the glycine site. GLYX-13 has been found to be a cognitive enhancer in learning models and is currently in clinical trials as an antidepressant.
GLYX-13 trifluoroacetate is a bain penetrable positive allosteric modulator of NMDA receptor acting via the glycine site; cognitive enhancer in learning models

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Propofol attenuates pancreatic cancer malignant potential via inhibition of NMDA receptor.
Chen X et al.
European Journal of Pharmacology, 795, 150-150 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service