Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0102

Sigma-Aldrich

Bac2A trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

L-Arginyl-L-leucyl-L-alanyl-L-arginyl-L-isoleucyl-L-valyl-L-valyl-L-isoleucyl-L-arginyl-L-valyl-L-alanyl- L-argininamide trifluoroacetate, RLARIVVIRVAR-amide trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H121N25O12 · xC2HF3O2
CAS Number:
Molecular Weight:
1420.80 (free base basis)
UNSPSC Code:
41116107

assay

≥98% (HPLC)

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Antimicrobial and immunomodulatory peptide
Bac2A is an antimicrobial and immunomodulatory peptide. Bac2A is a linear variant of bactenecin. Recent studies show that Bac2A is very effective against fungal pathogens, Fusarium oxysporum f. sp. vasinfectum and Fusarium graminearum. In these studies, Bac2A antifungal activity was not due to membrane destabilization but rather to interaction with intracellular targets.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service