Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0090

Sigma-Aldrich

PL415 trifluoroacetate salt hydrate

≥98% (HPLC)

Synonym(s):

Ac-Ser-Lys-Gly-Pya trifluoroacetate salt hydrate, AcSKGPya trifluoroacetate salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H37N5O7 · xC2HF3O2 · yH2O
Molecular Weight:
603.67 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

H2O: ≥10 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

O.OC(=O)C(F)(F)F.CC(=O)N[C@@H](CO)C(=O)N[C@@H](CCCCN)C(=O)NCC(=O)N[C@@H](Cc1cc2ccc3cccc4ccc(c1)c2c34)C(O)=O

InChI

1S/C32H37N5O7.C2HF3O2.H2O/c1-18(39)35-26(17-38)31(42)37-24(7-2-3-12-33)30(41)34-16-27(40)36-25(32(43)44)15-19-13-22-10-8-20-5-4-6-21-9-11-23(14-19)29(22)28(20)21;3-2(4,5)1(6)7;/h4-6,8-11,13-14,24-26,38H,2-3,7,12,15-17,33H2,1H3,(H,34,41)(H,35,39)(H,36,40)(H,37,42)(H,43,44);(H,6,7);1H2/t24-,25-,26-;;/m0../s1

InChI key

WRNVFQUPGFIUCI-YYBKPHAQSA-N

Biochem/physiol Actions

Fluorescent cleavage product of PL405, an endothelin converting enzyme-2 (ECE-2) fluorigenic substrate
PL415 is fluorescent cleavage product of PL405, an endothelin converting enzyme-2 (ECE-2) fluorigenic substrate. PL415 UV maximum absorption (λ max) is 342 nm (ε= 24 940L. mol-1.cm-1). PL415 is used to obtain calibration curve for kinetic studies utilizing PL405.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service